Assay Details
| Gene Symbol: RpL10Aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142153.3 | NP_650410.1 | 1 | 658 | 111 | |
| GenBank mRNA | AY113239.1 | - | 1 | 658 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RpL10Aa| Gene Symbol: RpL10Aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142153.3 | NP_650410.1 | 1 | 658 | 111 | |
| GenBank mRNA | AY113239.1 | - | 1 | 658 | 111 | |
Set Membership: |
|