Assay Details
| Gene Symbol: Set | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142181.3 | NP_650438.2 | 2 - 3 | 585 | 57 | |
| GenBank mRNA | AY118922.1 | - | 2 - 3 | 562 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Set| Gene Symbol: Set | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142181.3 | NP_650438.2 | 2 - 3 | 585 | 57 | |
| GenBank mRNA | AY118922.1 | - | 2 - 3 | 562 | 57 | |
Set Membership: |
|