Assay Details
| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142195.2 | NP_650452.1 | 2 - 3 | 1063 | 66 | |
| GenBank mRNA | BT100116.1 | - | 2 - 3 | 1034 | 66 | |
More Information
Set Membership: |
|

Atg4b
Genomic Map