Assay Details
| Gene Symbol: m-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275731.1 | NP_001262660.1 | 1 - 2 | 773 | 116 | |
| NM_142350.2 | NP_650607.1 | 1 - 2 | 773 | 116 | ||
| GenBank mRNA | BT133252.1 | - | 1 - 2 | 740 | 116 | |
More Information
Set Membership: |
|

m-cup
Genomic Map