Assay Details
| Gene Symbol: m-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142350.2 | NP_650607.1 | 4 - 5 | 2273 | 94 | |
| GenBank mRNA | BT133252.1 | - | 4 - 5 | 2240 | 94 | |
More Information
Set Membership: |
|

m-cup
Genomic Map