Assay Details
| Gene Symbol: m-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275731.1 | NP_001262660.1 | 5 - 6 | 2405 | 66 | |
| NM_142350.2 | NP_650607.1 | 5 - 6 | 2408 | 66 | ||
| GenBank mRNA | BT133252.1 | - | 5 - 6 | 2375 | 66 | |
More Information
Set Membership: |
|

m-cup
Genomic Map