Assay Details
| Gene Symbol: Gos28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142482.4 | NP_650739.2 | 3 - 4 | 654 | 97 | |
| GenBank mRNA | BT030790.1 | - | 3 - 4 | 650 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gos28| Gene Symbol: Gos28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142482.4 | NP_650739.2 | 3 - 4 | 654 | 97 | |
| GenBank mRNA | BT030790.1 | - | 3 - 4 | 650 | 97 | |
Set Membership: |
|