Assay Details
Gene Symbol: Nsun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142530.3 | NP_650787.1 | 3 - 4 | 935 | 67 | |
GenBank mRNA | AY069449.1 | - | 3 - 4 | 906 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
