Assay Details
| Gene Symbol: Pi3K92E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275841.1 | NP_001262770.1 | 5 - 6 | 3641 | 72 | |
| NM_142645.3 | NP_650902.1 | 5 - 6 | 3222 | 72 | ||
| NM_169910.2 | NP_732536.1 | 5 - 6 | 3149 | 72 | ||
| GenBank mRNA | BT003173.1 | - | 5 - 6 | 3191 | 72 | |
| Y09070.1 | - | 3137 | 72 | |||
More Information
Set Membership: |
|

Pi3K92E
Genomic Map