Assay Details
| Gene Symbol: RhoGAP93B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142682.2 | NP_650939.2 | 12 - 13 | 4174 | 76 | |
| GenBank mRNA | BT053685.1 | - | 12 - 13 | 4152 | 76 | |
More Information
Set Membership: |
|

RhoGAP93B
Genomic Map