Assay Details
| Gene Symbol: Idh3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170211.1 | NP_001163682.1 | 5 - 6 | 1231 | 53 | |
| NM_142743.3 | NP_651000.1 | 5 - 6 | 1033 | 53 | ||
| GenBank mRNA | BT004877.1 | - | 5 - 6 | 1198 | 53 | |
More Information
Set Membership: |
|

Idh3b
Genomic Map