Assay Details
| Gene Symbol: SppL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170269.2 | NP_001163740.1 | 5 - 6 | 1269 | 91 | |
| NM_001170270.2 | NP_001163741.1 | 4 - 5 | 1068 | 91 | ||
| NM_001300606.1 | NP_001287535.1 | 5 - 6 | 1207 | 91 | ||
| NM_143180.2 | NP_651437.1 | 4 - 5 | 1053 | 91 | ||
| NM_170244.2 | NP_733123.1 | 5 - 6 | 1254 | 91 | ||
| NM_170245.2 | NP_733124.1 | 4 - 5 | 810 | 91 | ||
| GenBank mRNA | BT003288.1 | - | 5 - 6 | 1239 | 91 | |
| BT132917.1 | - | 4 - 5 | 808 | 91 | ||
More Information
Set Membership: |
|

SppL
Genomic Map