Assay Details
| Gene Symbol: CG11837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143403.4 | NP_651660.1 | 2 - 3 | 876 | 120 | |
| GenBank mRNA | AY061429.1 | - | 2 - 3 | 858 | 120 | |
More Information
Set Membership: |
|

CG11837
Genomic Map