Assay Details
| Gene Symbol: eIF4H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276193.1 | NP_001263122.1 | 1 - 2 | 1064 | 54 | |
| NM_143563.3 | NP_651820.1 | 1 - 2 | 1064 | 54 | ||
| GenBank mRNA | BT003308.1 | - | 1 - 2 | 1063 | 54 | |
More Information
Set Membership: |
|

eIF4H2
Genomic Map