Assay Details
| Gene Symbol: CG15553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143584.4 | NP_651841.3 | 6 - 7 | 1253 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG15553| Gene Symbol: CG15553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143584.4 | NP_651841.3 | 6 - 7 | 1253 | 70 | |
Set Membership: |
|