Assay Details
| Gene Symbol: ATPsynC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260453.1 | NP_001247382.1 | 3 - 4 | 699 | 72 | |
| NM_001260454.1 | NP_001247383.1 | 2 - 3 | 319 | 72 | ||
| NM_001260455.2 | NP_001247384.1 | 2 - 3 | 316 | 72 | ||
| NM_143595.2 | NP_651852.1 | 3 - 4 | 408 | 72 | ||
| NM_170543.3 | NP_733422.1 | 2 - 3 | 316 | 72 | ||
| NM_170544.2 | NP_733423.1 | 3 - 4 | 763 | 72 | ||
| GenBank mRNA | BT011492.1 | - | 3 - 4 | 324 | 72 | |
| BT132846.1 | - | 2 - 3 | 319 | 72 | ||
| KX671606.1 | - | 2 - 3 | 255 | 72 | ||
More Information
Set Membership: |
|

ATPsynC
Genomic Map