Assay Details
| Gene Symbol: eEF1gamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202379.2 | NP_001189308.1 | 3 - 4 | 226 | 71 | |
| NM_143743.3 | NP_652000.1 | 2 - 3 | 199 | 71 | ||
| NM_170401.3 | NP_733280.1 | 3 - 4 | 222 | 71 | ||
More Information
Set Membership: |
|

eEF1gamma
Genomic Map