Assay Details
| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169562.2 | NP_731872.1 | 1 - 2 | 626 | 71 | |
| GenBank mRNA | AY113524.1 | - | 1 - 2 | 619 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ipp| Gene Symbol: Ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169562.2 | NP_731872.1 | 1 - 2 | 626 | 71 | |
| GenBank mRNA | AY113524.1 | - | 1 - 2 | 619 | 71 | |
Set Membership: |
|