Assay Details
| Gene Symbol: rec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275686.1 | NP_001262615.1 | 1 - 2 | 2274 | 106 | |
| NM_169674.2 | NP_732072.1 | 1 - 2 | 2274 | 106 | ||
More Information
Set Membership: |
|

rec
Genomic Map