Assay Details
| Gene Symbol: Mhcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043250.2 | NP_001036715.1 | 11 - 12 | 4202 | 106 | |
| NM_001043251.2 | NP_001036716.1 | 8 - 9 | 3929 | 106 | ||
| NM_169700.3 | NP_732108.2 | 11 - 12 | 4202 | 106 | ||
| NM_169701.3 | NP_732109.2 | 11 - 12 | 4202 | 106 | ||
| NM_170650.4 | NP_732110.2 | 2 - 3 | 343 | 106 | ||
| GenBank mRNA | BT001428.1 | - | 2 - 3 | 343 | 106 | |
More Information
Set Membership: |
|

Mhcl
Genomic Map