Assay Details
| Gene Symbol: Mhcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043250.2 | NP_001036715.1 | 15 - 16 | 6031 | 92 | |
| NM_001043251.2 | NP_001036716.1 | 12 - 13 | 5758 | 92 | ||
| NM_001202304.1 | NP_001189233.1 | 2 - 3 | 1264 | 92 | ||
| NM_001275694.1 | NP_001262623.1 | 5 - 6 | 3286 | 92 | ||
| NM_169700.3 | NP_732108.2 | 15 - 16 | 6031 | 92 | ||
| NM_169701.3 | NP_732109.2 | 15 - 16 | 6031 | 92 | ||
| NM_169703.1 | NP_732111.1 | 4 - 5 | 3349 | 92 | ||
| NM_170650.4 | NP_732110.2 | 6 - 7 | 2172 | 92 | ||
| GenBank mRNA | BT001428.1 | - | 6 - 7 | 2172 | 92 | |
More Information
Set Membership: |
|

Mhcl
Genomic Map