Assay Details
| Gene Symbol: Mhcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043250.2 | NP_001036715.1 | 16 - 17 | 6424 | 105 | |
| NM_001043251.2 | NP_001036716.1 | 13 - 14 | 6151 | 105 | ||
| NM_001202304.1 | NP_001189233.1 | 3 - 4 | 1657 | 105 | ||
| NM_001275694.1 | NP_001262623.1 | 6 - 7 | 3679 | 105 | ||
| NM_169700.3 | NP_732108.2 | 16 - 17 | 6424 | 105 | ||
| NM_169701.3 | NP_732109.2 | 16 - 17 | 6424 | 105 | ||
| NM_169703.1 | NP_732111.1 | 5 - 6 | 3742 | 105 | ||
| NM_170650.4 | NP_732110.2 | 7 - 8 | 2565 | 105 | ||
| GenBank mRNA | BT001428.1 | - | 7 - 8 | 2565 | 105 | |
More Information
Set Membership: |
|

Mhcl
Genomic Map