Assay Details
| Gene Symbol: Mhcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043250.2 | NP_001036715.1 | 2 - 3 | 616 | 78 | |
| NM_169700.3 | NP_732108.2 | 2 - 3 | 616 | 78 | ||
| NM_169701.3 | NP_732109.2 | 2 - 3 | 616 | 78 | ||
More Information
Set Membership: |
|

Mhcl
Genomic Map