Assay Details
| Gene Symbol: Mhcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043250.2 | NP_001036715.1 | 9 - 10 | 3869 | 58 | |
| NM_001043251.2 | NP_001036716.1 | 6 - 7 | 3596 | 58 | ||
| NM_169700.3 | NP_732108.2 | 9 - 10 | 3869 | 58 | ||
| NM_169701.3 | NP_732109.2 | 9 - 10 | 3869 | 58 | ||
More Information
Set Membership: |
|

Mhcl
Genomic Map