Assay Details
| Gene Symbol: Ino80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169854.2 | NP_732413.1 | 6 - 7 | 3465 | 91 | |
| GenBank mRNA | BT133394.1 | - | 6 - 7 | 3365 | 91 | |
More Information
Set Membership: |
|

Ino80
Genomic Map