Assay Details
| Gene Symbol: Ino80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169854.2 | NP_732413.1 | 9 - 10 | 4055 | 62 | |
| GenBank mRNA | BT133394.1 | - | 9 - 10 | 3955 | 62 | |
More Information
Set Membership: |
|

Ino80
Genomic Map