Assay Details
| Gene Symbol: SNF4Agamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043271.3 | NP_001036736.2 | 8 - 9 | 1273 | 80 | |
| NM_001043272.2 | NP_001036737.1 | 8 - 9 | 2183 | 80 | ||
| NM_001043273.3 | NP_001036738.1 | 8 - 9 | 2730 | 80 | ||
| NM_001043274.2 | NP_001036739.1 | 5 - 6 | 1164 | 80 | ||
| NM_001104384.3 | NP_001097854.1 | 8 - 9 | 1243 | 80 | ||
| NM_001104385.2 | NP_001097855.1 | 8 - 9 | 1579 | 80 | ||
| NM_001170198.2 | NP_001163669.1 | 8 - 9 | 2247 | 80 | ||
| NM_001170199.2 | NP_001163670.1 | 8 - 9 | 2298 | 80 | ||
| NM_001170200.2 | NP_001163671.1 | 12 - 13 | 3765 | 80 | ||
| NM_001170201.2 | NP_001163672.1 | 12 - 13 | 3647 | 80 | ||
| NM_080509.3 | NP_536757.2 | 12 - 13 | 2498 | 80 | ||
| NM_169946.2 | NP_732594.1 | 12 - 13 | 2669 | 80 | ||
| NM_169947.3 | NP_732598.1 | 12 - 13 | 3824 | 80 | ||
| NM_169948.3 | NP_732599.1 | 8 - 9 | 2252 | 80 | ||
| NM_169949.3 | NP_732600.1 | 8 - 9 | 2363 | 80 | ||
| NM_169950.3 | NP_732601.1 | 5 - 6 | 1841 | 80 | ||
| GenBank mRNA | AY070541.1 | - | 12 - 13 | 3801 | 80 | |
| AY084138.1 | - | 8 - 9 | 2250 | 80 | ||
| AY166752.1 | - | 12 - 13 | 3808 | 80 | ||
| AY166754.1 | - | 5 - 6 | 1896 | 80 | ||
| BT003249.1 | - | 5 - 6 | 1819 | 80 | ||
| BT003626.1 | - | 2498 | 80 | |||
| BT010233.1 | - | 12 - 13 | 2667 | 80 | ||
| BT029676.1 | - | 8 - 9 | 1273 | 80 | ||
| BT029715.1 | - | 8 - 9 | 1243 | 80 | ||
| BT120301.1 | - | 12 - 13 | 2517 | 80 | ||
More Information
Set Membership: |
|

SNF4Agamma
Genomic Map