Assay Details
| Gene Symbol: SNF4Agamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043272.2 | NP_001036737.1 | 2 - 3 | 686 | 127 | |
| NM_001043273.3 | NP_001036738.1 | 2 - 3 | 1233 | 127 | ||
| NM_001170198.2 | NP_001163669.1 | 2 - 3 | 750 | 127 | ||
| NM_001170199.2 | NP_001163670.1 | 2 - 3 | 801 | 127 | ||
| NM_169948.3 | NP_732599.1 | 2 - 3 | 755 | 127 | ||
| NM_169949.3 | NP_732600.1 | 2 - 3 | 866 | 127 | ||
| GenBank mRNA | AY084138.1 | - | 2 - 3 | 753 | 127 | |
More Information
Set Membership: |
|

SNF4Agamma
Genomic Map