Assay Details
| Gene Symbol: LpR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104456.2 | NP_001097926.1 | 3 - 4 | 932 | 59 | |
| NM_001104457.2 | NP_001097927.1 | 3 - 4 | 932 | 59 | ||
| NM_001104458.2 | NP_001097928.1 | 3 - 4 | 932 | 59 | ||
| NM_001104460.2 | NP_001097930.1 | 3 - 4 | 932 | 59 | ||
| NM_170239.4 | NP_733118.3 | 3 - 4 | 932 | 59 | ||
| GenBank mRNA | BT010074.1 | - | 3 - 4 | 910 | 59 | |
| BT011437.1 | - | 3 - 4 | 933 | 59 | ||
More Information
Set Membership: |
|

LpR2
Genomic Map