Assay Details
| Gene Symbol: LpR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104456.2 | NP_001097926.1 | 7 - 8 | 2802 | 64 | |
| NM_001104458.2 | NP_001097928.1 | 7 - 8 | 2802 | 64 | ||
| NM_001104459.2 | NP_001097929.1 | 6 - 7 | 2261 | 64 | ||
| NM_001104460.2 | NP_001097930.1 | 7 - 8 | 2799 | 64 | ||
| NM_001276041.1 | NP_001262970.1 | 6 - 7 | 2250 | 64 | ||
| NM_001276042.1 | NP_001262971.1 | 6 - 7 | 2250 | 64 | ||
| NM_170239.4 | NP_733118.3 | 7 - 8 | 2799 | 64 | ||
| NM_176565.2 | NP_788742.1 | 6 - 7 | 2194 | 64 | ||
| GenBank mRNA | BT010074.1 | - | 7 - 8 | 2780 | 64 | |
| BT010094.1 | - | 6 - 7 | 2172 | 64 | ||
More Information
Set Membership: |
|

LpR2
Genomic Map