Assay Details
| Gene Symbol: LpR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104456.2 | NP_001097926.1 | 8 - 9 | 2987 | 79 | |
| NM_001276042.1 | NP_001262971.1 | 7 - 8 | 2435 | 79 | ||
| NM_170239.4 | NP_733118.3 | 8 - 9 | 2984 | 79 | ||
| NM_176565.2 | NP_788742.1 | 7 - 8 | 2379 | 79 | ||
More Information
Set Membership: |
|

LpR2
Genomic Map