Assay Details
| Gene Symbol: AstA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300582.1 | NP_001287511.1 | 2 - 3 | 659 | 68 | |
| NM_079765.3 | NP_524489.2 | 2 - 3 | 662 | 68 | ||
| GenBank mRNA | AY071110.1 | - | 2 - 3 | 651 | 68 | |
| KX531458.1 | - | 1 - 2 | 387 | 68 | ||
More Information
Set Membership: |
|

AstA
Genomic Map