Assay Details
| Gene Symbol: His2Av | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276068.1 | NP_001262997.1 | 2 - 3 | 251 | 78 | |
| NM_079795.3 | NP_524519.1 | 2 - 3 | 251 | 78 | ||
| GenBank mRNA | AY118910.1 | - | 2 - 3 | 224 | 78 | |
| X07485.1 | - | 236 | 78 | |||
More Information
Set Membership: |
|

His2Av
Genomic Map