Assay Details
| Gene Symbol: Tsp97E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144643.3 | NP_001138115.2 | 2 - 3 | 333 | 70 | |
| NM_001276073.1 | NP_001263002.1 | 2 - 3 | 333 | 70 | ||
| NM_001300621.1 | NP_001287550.1 | 2 - 3 | 333 | 70 | ||
| NM_079800.3 | NP_524524.1 | 2 - 3 | 333 | 70 | ||
| NM_170297.3 | NP_733176.2 | 2 - 3 | 333 | 70 | ||
| GenBank mRNA | AF274020.1 | - | 2 - 3 | 341 | 70 | |
| AY060360.1 | - | 2 - 3 | 341 | 70 | ||
More Information
Set Membership: |
|

Tsp97E
Genomic Map