Assay Details
| Gene Symbol: AstA-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260423.2 | NP_001247352.1 | 2 - 3 | 1576 | 85 | |
| NM_001276113.1 | NP_001263042.1 | 2 - 3 | 1576 | 85 | ||
| NM_079820.3 | NP_524544.1 | 2 - 3 | 1576 | 85 | ||
| GenBank mRNA | AF253526.1 | - | 2 - 3 | 927 | 85 | |
| BT003199.1 | - | 2 - 3 | 1577 | 85 | ||
| BT125007.1 | - | 2 - 3 | 1557 | 85 | ||
More Information
Set Membership: |
|

AstA-R2
Genomic Map