Assay Details
| Gene Symbol: sima | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276171.1 | NP_001263100.1 | 5 - 6 | 3037 | 88 | |
| NM_001300670.1 | NP_001287599.1 | 11 - 12 | 4311 | 88 | ||
| NM_079845.4 | NP_524584.2 | 11 - 12 | 4511 | 88 | ||
More Information
Set Membership: |
|

sima
Genomic Map