Assay Details
| Gene Symbol: sima | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144657.3 | NP_001138129.1 | 6 - 7 | 1737 | 86 | |
| NM_001300670.1 | NP_001287599.1 | 6 - 7 | 1537 | 86 | ||
| NM_079845.4 | NP_524584.2 | 6 - 7 | 1737 | 86 | ||
More Information
Set Membership: |
|

sima
Genomic Map