Assay Details
| Gene Symbol: sima | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300670.1 | NP_001287599.1 | 7 - 8 | 1677 | 76 | |
| NM_079845.4 | NP_524584.2 | 7 - 8 | 1877 | 76 | ||
More Information
Set Membership: |
|

sima
Genomic Map