Assay Details
| Gene Symbol: Med | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079871.4 | NP_524610.1 | 1 - 2 | 700 | 69 | |
| NM_170559.3 | NP_733438.1 | 1 - 2 | 700 | 69 | ||
| NM_170560.2 | NP_733439.2 | 1 - 2 | 700 | 69 | ||
| GenBank mRNA | AF027729.1 | - | 1 - 2 | 681 | 69 | |
| AF041439.1 | - | 1 - 2 | 275 | 69 | ||
| AY069496.1 | - | 1 - 2 | 675 | 69 | ||
More Information
Set Membership: |
|

Med
Genomic Map