Assay Details
| Gene Symbol: Med | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079871.4 | NP_524610.1 | 7 - 8 | 2616 | 84 | |
| NM_170559.3 | NP_733438.1 | 6 - 7 | 2394 | 84 | ||
| NM_170560.2 | NP_733439.2 | 7 - 8 | 2616 | 84 | ||
| GenBank mRNA | AF027729.1 | - | 7 - 8 | 2597 | 84 | |
| AF041439.1 | - | 7 - 8 | 2191 | 84 | ||
| AY069496.1 | - | 7 - 8 | 2591 | 84 | ||
More Information
Set Membership: |
|

Med
Genomic Map