Assay Details
| Gene Symbol: eEF1alpha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079872.4 | NP_524611.1 | 3 - 4 | 1006 | 63 | |
| NM_170570.1 | NP_733449.1 | 3 - 4 | 994 | 63 | ||
| NM_206592.1 | NP_996315.1 | 3 - 4 | 1003 | 63 | ||
| NM_206593.1 | NP_996316.1 | 3 - 4 | 991 | 63 | ||
| GenBank mRNA | BT015202.1 | - | 3 - 4 | 1006 | 63 | |
More Information
Set Membership: |
|

eEF1alpha2
Genomic Map