Assay Details
| Gene Symbol: lds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080111.4 | NP_524850.2 | 1 - 2 | 168 | 59 | |
| GenBank mRNA | BT150462.1 | - | 1 - 2 | 166 | 59 | |
| BT150466.1 | - | 1 - 2 | 166 | 59 | ||
More Information
Set Membership: |
|

lds
Genomic Map