Assay Details
| Gene Symbol: lds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080111.4 | NP_524850.2 | 4 - 5 | 2516 | 90 | |
| GenBank mRNA | BT150462.1 | - | 4 - 5 | 2514 | 90 | |
| BT150466.1 | - | 4 - 5 | 2514 | 90 | ||
More Information
Set Membership: |
|

lds
Genomic Map