Assay Details
| Gene Symbol: cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104373.2 | NP_001097843.1 | 9 - 10 | 5209 | 60 | |
| NM_001260273.2 | NP_001247202.1 | 9 - 10 | 3919 | 60 | ||
| NM_001260274.2 | NP_001247203.1 | 15 - 16 | 6437 | 60 | ||
| NM_001260275.2 | NP_001247204.2 | 9 - 10 | 3707 | 60 | ||
| NM_001275826.1 | NP_001262755.1 | 14 - 15 | 7159 | 60 | ||
| NM_080253.4 | NP_524992.1 | 9 - 10 | 3922 | 60 | ||
| GenBank mRNA | AJ252268.1 | - | 9 - 10 | 3900 | 60 | |
| BT044183.1 | - | 9 - 10 | 3901 | 60 | ||
More Information
Set Membership: |
|

cic
Genomic Map