Assay Details
| Gene Symbol: Med | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170559.3 | NP_733438.1 | 3 - 4 | 1323 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med| Gene Symbol: Med | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170559.3 | NP_733438.1 | 3 - 4 | 1323 | 87 | |
Set Membership: |
|