Assay Details
| Gene Symbol: RpL34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202366.1 | NP_001189295.1 | 2 - 3 | 101 | 132 | |
| NM_143203.3 | NP_651460.2 | 2 - 3 | 104 | 132 | ||
| NM_170257.3 | NP_733136.1 | 2 - 3 | 119 | 132 | ||
| GenBank mRNA | BT032652.1 | - | 2 - 3 | 100 | 132 | |
| KX531929.1 | - | 1 - 2 | 61 | 132 | ||
More Information
Set Membership: |
|

RpL34a
Genomic Map