Assay Details
| Gene Symbol: mRpS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078749.4 | NP_523473.2 | 2 | 829 | 113 | |
| GenBank mRNA | AY118630.1 | - | 2 | 829 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpS2| Gene Symbol: mRpS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078749.4 | NP_523473.2 | 2 | 829 | 113 | |
| GenBank mRNA | AY118630.1 | - | 2 | 829 | 113 | |
Set Membership: |
|