Assay Details
| Gene Symbol: Pk34A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135759.2 | NP_609603.1 | 2 | 1130 | 68 | |
| GenBank mRNA | BT120381.1 | - | 1 | 1061 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pk34A| Gene Symbol: Pk34A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135759.2 | NP_609603.1 | 2 | 1130 | 68 | |
| GenBank mRNA | BT120381.1 | - | 1 | 1061 | 68 | |
Set Membership: |
|