Assay Details
| Gene Symbol: ytr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299856.1 | NP_001286785.1 | 4 | 1916 | 108 | |
| NM_137995.3 | NP_611839.1 | 3 | 1923 | 108 | ||
| GenBank mRNA | AY061143.1 | - | 3 | 1946 | 108 | |
More Information
Set Membership: |
|

ytr
Genomic Map