Assay Details
| Gene Symbol: Cpr60D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299924.1 | NP_001286853.1 | 2 | 362 | 106 | |
| NM_166679.2 | NP_726468.1 | 2 | 448 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr60D| Gene Symbol: Cpr60D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299924.1 | NP_001286853.1 | 2 | 362 | 106 | |
| NM_166679.2 | NP_726468.1 | 2 | 448 | 106 | ||
Set Membership: |
|